Assay Details
Gene Symbol: TREX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO624885.2 | - | 4 - 5 | 578 | 98 |
Gene Symbol: ATRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018067022.1 | XP_017922511.1 | 9 - 10 | 1979 | 98 | |
XM_018067024.1 | XP_017922513.1 | 9 - 10 | 1979 | 98 | ||
GenBank mRNA | JO591461.2 | - | 4 - 5 | 717 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
