Assay Details
| Gene Symbol: GLRX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041489.1 | XP_017896978.1 | 10 - 11 | 986 | 82 | |
| GenBank mRNA | JO622642.2 | - | 1 - 2 | 289 | 82 | |
More Information
Set Membership: |
|

GLRX3
Genomic Map