Assay Details
| Gene Symbol: LOC102183767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042672.1 | XP_017898161.1 | 14 - 15 | 1707 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102183767| Gene Symbol: LOC102183767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042672.1 | XP_017898161.1 | 14 - 15 | 1707 | 106 | |
Set Membership: |
|