Assay Details
| Gene Symbol: CLEC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680854.3 | XP_005680911.1 | 5 - 6 | 655 | 76 | |
| GenBank mRNA | JO409292.2 | - | 5 - 6 | 626 | 76 | |
More Information
Set Membership: |
|

CLEC1B
Genomic Map