Assay Details
| Gene Symbol: CCDC77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680998.3 | XP_005681055.1 | 10 - 11 | 1305 | 59 | |
| XM_005681000.3 | XP_005681057.1 | 10 - 11 | 1257 | 59 | ||
More Information
Set Membership: |
|

CCDC77
Genomic Map