Assay Details
Gene Symbol: ZNF461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692333.3 | XP_005692390.2 | 4 - 5 | 572 | 64 | |
XM_013971156.2 | XP_013826610.2 | 4 - 5 | 594 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
