Assay Details
| Gene Symbol: SMARCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065037.1 | XP_017920526.1 | 8 - 9 | 1332 | 71 | |
| XM_018065038.1 | XP_017920527.1 | 8 - 9 | 1332 | 71 | ||
| XM_018065039.1 | XP_017920528.1 | 7 - 8 | 1054 | 71 | ||
More Information
Set Membership: |
|

SMARCE1
Genomic Map