Assay Details
| Gene Symbol: SUMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695707.2 | XP_005695764.1 | 8 - 9 | 1027 | 66 | |
| XM_005695710.2 | XP_005695767.1 | 7 - 8 | 913 | 66 | ||
| GenBank mRNA | JO429501.2 | - | 8 - 9 | 1042 | 66 | |
More Information
Set Membership: |
|

SUMF1
Genomic Map