Assay Details
| Gene Symbol: DCUN1D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697571.3 | XP_005697628.1 | 4 - 5 | 918 | 75 | |
| XM_018040910.1 | XP_017896399.1 | 4 - 5 | 908 | 75 | ||
| XM_018040911.1 | XP_017896400.1 | 3 - 4 | 832 | 75 | ||
| XM_018040912.1 | XP_017896401.1 | 2 - 3 | 740 | 75 | ||
| XM_018040913.1 | XP_017896402.1 | 3 - 4 | 871 | 75 | ||
| XM_018040914.1 | XP_017896403.1 | 4 - 5 | 835 | 75 | ||
| XM_018040915.1 | XP_017896404.1 | 2 - 3 | 667 | 75 | ||
More Information
Set Membership: |
|

DCUN1D3
Genomic Map