Assay Details
| Gene Symbol: LOC102188999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697765.3 | XP_005697822.2 | 11 - 12 | 1548 | 91 | |
| GenBank mRNA | JO426248.1 | - | 11 - 12 | 1622 | 91 | |
| JO428960.2 | - | 11 - 12 | 1562 | 91 | ||
More Information
Set Membership: |
|

LOC102188999
Genomic Map