Assay Details
Gene Symbol: BAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042790.1 | XP_017898279.1 | 3 - 4 | 495 | 63 | |
XM_018042791.1 | XP_017898280.1 | 3 - 4 | 671 | 63 | ||
GenBank mRNA | JO440936.2 | - | 3 - 4 | 483 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
