Assay Details
| Gene Symbol: SPATA31C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013966085.2 | XP_013821539.2 | 6 - 7 | 3654 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPATA31C2| Gene Symbol: SPATA31C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013966085.2 | XP_013821539.2 | 6 - 7 | 3654 | 107 | |
Set Membership: |
|