Assay Details
| Gene Symbol: CD8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055352.1 | XP_017910841.1 | 4 - 5 | 631 | 50 | |
| GenBank mRNA | JO326107.2 | - | 2 - 3 | 733 | 50 | |
More Information
Set Membership: |
|

CD8B
Genomic Map