Assay Details
Gene Symbol: AVPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045070.1 | XP_017900559.1 | 2 - 3 | 1380 | 118 | |
XM_018045071.1 | XP_017900560.1 | 3 - 4 | 1094 | 118 | ||
GenBank mRNA | JO459081.2 | - | 1 - 2 | 385 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
