Assay Details
| Gene Symbol: LOC102169149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046133.1 | XP_017901622.1 | 2 - 3 | 305 | 83 | |
| XM_018046134.1 | XP_017901623.1 | 2 - 3 | 386 | 83 | ||
| GenBank mRNA | JO339655.2 | - | 2 - 3 | 310 | 83 | |
| KA337931.1 | - | 1 - 2 | 268 | 83 | ||
More Information
Set Membership: |
|

LOC102169149
Genomic Map