Assay Details
| Gene Symbol: GPRC5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680813.3 | XP_005680870.1 | 2 - 3 | 1163 | 66 | |
| GenBank mRNA | JO349369.2 | - | 1 - 2 | 923 | 66 | |
More Information
Set Membership: |
|

GPRC5A
Genomic Map