Assay Details
| Gene Symbol: CALHM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684537.3 | XP_005684594.1 | 1 - 2 | 618 | 66 | |
| GenBank mRNA | KA313023.1 | - | 2 - 3 | 1228 | 66 | |
More Information
Set Membership: |
|

CALHM5
Genomic Map