Assay Details
Gene Symbol: IPCEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053520.1 | XP_017909009.1 | 2 - 3 | 316 | 76 | |
GenBank mRNA | JT704601.1 | - | 10 - 11 | 1577 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
