Assay Details
Gene Symbol: CEBPZOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686493.3 | XP_005686550.1 | 5 - 6 | 451 | 91 | |
XM_013967556.2 | XP_013823010.1 | 4 - 5 | 318 | 91 | ||
XM_018055127.1 | XP_017910616.1 | 5 - 6 | 380 | 91 | ||
XM_018055128.1 | XP_017910617.1 | 5 - 6 | 424 | 91 | ||
XM_018055129.1 | XP_017910618.1 | 4 - 5 | 340 | 91 | ||
GenBank mRNA | KA350448.1 | - | 1 - 2 | 180 | 91 |
Gene Symbol: LOC108633904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040888.1 | XP_017896377.1 | 1 | 760 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
