Assay Details
| Gene Symbol: KBTBD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687411.3 | XP_005687468.2 | 1 | 2062 | 72 | |
| GenBank mRNA | JT699880.1 | - | 1 | 312 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KBTBD7| Gene Symbol: KBTBD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687411.3 | XP_005687468.2 | 1 | 2062 | 72 | |
| GenBank mRNA | JT699880.1 | - | 1 | 312 | 72 | |
Set Membership: |
|