Assay Details
Gene Symbol: SPART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687509.3 | XP_005687566.2 | 6 - 7 | 1620 | 64 | |
XM_018056658.1 | XP_017912147.1 | 6 - 7 | 1682 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
