Assay Details
| Gene Symbol: C16H1orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690559.1 | XP_005690616.1 | 4 - 5 | 432 | 104 | |
| XM_013970082.1 | XP_013825536.1 | 3 - 4 | 243 | 104 | ||
More Information
Set Membership: |
|

C16H1orf100
Genomic Map