Assay Details
| Gene Symbol: PDGFC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691309.3 | XP_005691366.2 | 5 - 6 | 1098 | 58 | |
| XM_018061475.1 | XP_017916964.1 | 5 - 6 | 1151 | 58 | ||
More Information
Set Membership: |
|

PDGFC
Genomic Map