Assay Details
| Gene Symbol: MRPL58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063840.1 | XP_017919329.1 | 5 - 6 | 578 | 64 | |
| XM_018063841.1 | XP_017919330.1 | 5 - 6 | 486 | 64 | ||
| GenBank mRNA | JO577132.1 | - | 1 - 2 | 386 | 64 | |
| JO577133.1 | - | 1 - 2 | 386 | 64 | ||
More Information
Set Membership: |
|

MRPL58
Genomic Map