Assay Details
Gene Symbol: LOC108633235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043551.1 | XP_017899040.1 | 1 - 2 | 325 | 85 |
Gene Symbol: LOC102175469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043600.1 | XP_017899089.1 | 1 - 2 | 325 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
