Assay Details
| Gene Symbol: BLZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005709424.3 | XP_005709481.2 | 5 - 6 | 1185 | 73 | |
| XM_018060281.1 | XP_017915770.1 | 5 - 6 | 1185 | 73 | ||
| GenBank mRNA | JO403370.2 | - | 2 - 3 | 492 | 73 | |
| JO585192.2 | - | 2 - 3 | 693 | 73 | ||
More Information
Set Membership: |
|

BLZF1
Genomic Map