Assay Details
| Gene Symbol: PDCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963098.2 | XP_013818552.2 | 2 - 3 | 455 | 102 | |
| XM_018044475.1 | XP_017899964.1 | 2 - 3 | 455 | 102 | ||
| GenBank mRNA | JO608403.2 | - | 2 - 3 | 477 | 102 | |
More Information
Set Membership: |
|

PDCD1
Genomic Map