Assay Details
Gene Symbol: BLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051835.1 | XP_017907324.1 | 4 - 5 | 404 | 59 | |
XM_018051836.1 | XP_017907325.1 | 4 - 5 | 340 | 59 | ||
GenBank mRNA | JT715138.1 | - | 9 - 10 | 1970 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
