Assay Details
Gene Symbol: NDUFAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058244.1 | XP_017913733.1 | 2 - 3 | 320 | 73 | |
XM_018058245.1 | XP_017913734.1 | 2 - 3 | 320 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
