Assay Details
| Gene Symbol: CCNQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044785.1 | XP_017900274.1 | 3 - 4 | 585 | 64 | |
| XM_018044786.1 | XP_017900275.1 | 3 - 4 | 582 | 64 | ||
| XM_018044787.1 | XP_017900276.1 | 3 - 4 | 582 | 64 | ||
| XM_018044788.1 | XP_017900277.1 | 4 - 5 | 580 | 64 | ||
| XM_018044789.1 | XP_017900278.1 | 3 - 4 | 438 | 64 | ||
| XM_018044790.1 | XP_017900279.1 | 3 - 4 | 421 | 64 | ||
More Information
Set Membership: |
|

CCNQ
Genomic Map