Assay Details
| Gene Symbol: APOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675093.3 | XP_005675150.1 | 4 - 5 | 647 | 62 | |
| GenBank mRNA | JO346885.2 | - | 1 - 2 | 574 | 62 | |
| JO405496.2 | - | 1 - 2 | 510 | 62 | ||
| JO439300.2 | - | 1 - 2 | 533 | 62 | ||
| JO504364.2 | - | 4 - 5 | 602 | 62 | ||
| JO537085.2 | - | 1 - 2 | 815 | 62 | ||
| JO562276.2 | - | 1 - 2 | 524 | 62 | ||
| JO595749.2 | - | 4 - 5 | 449 | 62 | ||
| JO595750.2 | - | 5 - 6 | 544 | 62 | ||
| JP350938.2 | - | 1 - 2 | 815 | 62 | ||
| JT763244.1 | - | 4 - 5 | 468 | 62 | ||
| KA315257.1 | - | 4 - 5 | 699 | 62 | ||
More Information
Set Membership: |
|

APOD
Genomic Map