Assay Details
Gene Symbol: CPEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049737.1 | XP_017905226.1 | 10 - 11 | 2717 | 61 | |
XM_018049738.1 | XP_017905227.1 | 9 - 10 | 2693 | 61 | ||
XM_018049739.1 | XP_017905228.1 | 8 - 9 | 2976 | 61 | ||
XM_018049740.1 | XP_017905229.1 | 9 - 10 | 2627 | 61 | ||
XM_018049741.1 | XP_017905230.1 | 8 - 9 | 2603 | 61 | ||
XM_018049742.1 | XP_017905231.1 | 7 - 8 | 2887 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
