Assay Details
| Gene Symbol: CCL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684094.3 | XP_005684151.1 | 1 - 2 | 259 | 91 | |
| XM_005684095.3 | XP_005684152.1 | 1 - 2 | 274 | 91 | ||
More Information
Set Membership: |
|

CCL19
Genomic Map