Assay Details
| Gene Symbol: LOC100861326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696862.3 | XP_005696919.2 | 1 - 2 | 1373 | 83 | |
| GenBank mRNA | JI861641.1 | - | 1 - 2 | 925 | 83 | |
More Information
Set Membership: |
|

LOC100861326
Genomic Map