Assay Details
Gene Symbol: MAP1S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051398.1 | XP_017906887.1 | 3 - 4 | 343 | 59 | |
GenBank mRNA | JT663811.1 | - | 3 - 4 | 2437 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
