Assay Details
Gene Symbol: LXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013965102.2 | XP_013820556.1 | 3 - 4 | 403 | 69 | |
GenBank mRNA | JI861855.1 | - | 3 - 4 | 364 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
