Assay Details
Gene Symbol: H6PD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060387.1 | XP_017915876.1 | 2 - 3 | 856 | 64 | |
XM_018060388.1 | XP_017915877.1 | 2 - 3 | 1048 | 64 | ||
GenBank mRNA | JO425043.1 | - | 3 - 4 | 3278 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
