Assay Details
| Gene Symbol: NAPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039498.1 | XP_017894987.1 | 5 - 6 | 745 | 68 | |
| XM_018039499.1 | XP_017894988.1 | 5 - 6 | 721 | 68 | ||
| XM_018039500.1 | XP_017894989.1 | 5 - 6 | 745 | 68 | ||
| GenBank mRNA | JO426882.1 | - | 5 - 6 | 348 | 68 | |
| JO598422.2 | - | 2 - 3 | 277 | 68 | ||
More Information
Set Membership: |
|

NAPG
Genomic Map