Assay Details
Gene Symbol: FAM20C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040598.1 | XP_017896087.1 | 6 - 7 | 2257 | 58 | |
GenBank mRNA | JO348171.2 | - | 6 - 7 | 953 | 58 | |
JO548249.2 | - | 6 - 7 | 898 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
