Assay Details
Gene Symbol: MFHAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041952.1 | XP_017897441.1 | 2 - 3 | 3232 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
