Assay Details
| Gene Symbol: LOC102171473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013976013.2 | XP_013831467.2 | 2 - 3 | 363 | 63 | |
| GenBank mRNA | JO352480.2 | - | 4 - 5 | 725 | 63 | |
| KA347150.1 | - | 4 - 5 | 2550 | 63 | ||
More Information
Set Membership: |
|

LOC102171473
Genomic Map