Assay Details
| Gene Symbol: CNIH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043603.1 | XP_017899092.1 | 3 - 4 | 455 | 83 | |
| GenBank mRNA | JO644625.2 | - | 3 - 4 | 489 | 83 | |
| JO644626.2 | - | 3 - 4 | 493 | 83 | ||
More Information
Set Membership: |
|

CNIH2
Genomic Map