Assay Details
| Gene Symbol: LOC102180115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO420746.2 | - | 900 | 68 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102180115| Gene Symbol: LOC102180115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO420746.2 | - | 900 | 68 | ||