Assay Details
| Gene Symbol: CEP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675072.3 | XP_005675129.1 | 2 - 3 | 324 | 80 | |
| XM_005675073.3 | XP_005675130.1 | 2 - 3 | 322 | 80 | ||
| GenBank mRNA | JO442444.2 | - | 1 - 2 | 647 | 80 | |
| JO601585.2 | - | 2 - 3 | 341 | 80 | ||
| JO642595.2 | - | 1 - 2 | 188 | 80 | ||
| KA306065.1 | - | 1 - 2 | 419 | 80 | ||
More Information
Set Membership: |
|

CEP19
Genomic Map