Assay Details
| Gene Symbol: HAO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677786.3 | XP_005677843.1 | 5 - 6 | 884 | 55 | |
| XM_005677789.3 | XP_005677846.1 | 5 - 6 | 902 | 55 | ||
| XM_013962496.2 | XP_013817950.1 | 6 - 7 | 986 | 55 | ||
| XM_013962497.2 | XP_013817951.1 | 7 - 8 | 1017 | 55 | ||
| GenBank mRNA | JO420799.2 | - | 3 - 4 | 1561 | 55 | |
More Information
Set Membership: |
|

HAO2
Genomic Map