Assay Details
Gene Symbol: BEND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043975.1 | XP_017899464.1 | 5 - 6 | 1239 | 86 | |
XM_018043979.1 | XP_017899468.1 | 6 - 7 | 1148 | 86 | ||
XM_018043986.1 | XP_017899475.1 | 5 - 6 | 1321 | 86 | ||
XM_018043991.1 | XP_017899480.1 | 4 - 5 | 864 | 86 | ||
GenBank mRNA | JO431058.2 | - | 5 - 6 | 1385 | 86 | |
JT664665.1 | - | 6 - 7 | 1412 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
