Assay Details
Gene Symbol: UTP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678674.3 | XP_005678731.1 | 1 - 2 | 164 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
