Assay Details
| Gene Symbol: OCEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682133.3 | XP_005682190.1 | 4 - 5 | 801 | 63 | |
| XM_018051368.1 | XP_017906857.1 | 5 - 6 | 881 | 63 | ||
More Information
Set Membership: |
|

OCEL1
Genomic Map