Assay Details
| Gene Symbol: PPM1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055501.1 | XP_017910990.1 | 6 - 7 | 1227 | 55 | |
| GenBank mRNA | JO339739.2 | - | 4 - 5 | 1062 | 55 | |
More Information
Set Membership: |
|

PPM1G
Genomic Map