Assay Details
| Gene Symbol: LOC102180330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689128.2 | XP_005689185.1 | 3 - 4 | 458 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102180330| Gene Symbol: LOC102180330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689128.2 | XP_005689185.1 | 3 - 4 | 458 | 87 | |
Set Membership: |
|